siRNA Details
| Gene Symbol: EDN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001956.4 | NP_001947.1 | 4, 5 | 523 | ||
| NR_126098.1 | 4, 5 | 432 | ||||
| GenBank mRNA | BC034393.1 | - | 4, 5 | 466 | 5185394 | |
| BI761655.1 | - | 4, 5 | 421 | |||
| M65199.1 | - | 4, 5 | 511 | |||
| S63835.1 | - | 2, 3 | 136 | |||
| S63837.1 | - | 3, 4 | 259 | |||
| S63838.1 | - | 4, 5 | 311 | |||
| X55177.1 | - | 4, 5 | 499 | |||

EDN2
Genomic Map