siRNA Details
| Gene Symbol: CPNE8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153634.2 | NP_705898.1 | 11 | 867 | ||
| XM_011537951.2 | XP_011536253.1 | 11 | 3842 | |||
| XM_011537952.2 | XP_011536254.1 | 11 | 3842 | |||
| XM_017018851.1 | XP_016874340.1 | 12 | 1840 | |||
| XM_017018852.1 | XP_016874341.1 | 11 | 813 | |||
| GenBank mRNA | AK098593.1 | - | 11 | 797 | ||
| AK307908.1 | - | 11 | 840 | |||
| AY177785.1 | - | 11 | 783 | |||
| BC068040.1 | - | 11 | 831 | |||
| BC110578.1 | - | 11 | 858 | 40007744 | ||
| BX538055.1 | - | 11 | 881 | |||

CPNE8
Genomic Map