siRNA Details
| Gene Symbol: FBXL14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152441.2 | NP_689654.1 | 1 | 1201 | ||
| XM_017018875.1 | XP_016874364.1 | 1 | 1225 | |||
| XM_017018876.1 | XP_016874365.1 | 1 | 1225 | |||
| GenBank mRNA | BC028132.1 | - | 1 | 1201 | 5205284 | |

FBXL14
Genomic Map