siRNA Details
| Gene Symbol: FAM109A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177996.1 | NP_001171467.1 | 4 | 1639 | ||
| NM_001177997.1 | NP_001171468.1 | 3 | 1532 | |||
| NM_144671.4 | NP_653272.2 | 3 | 1551 | |||
| GenBank mRNA | AK056918.1 | - | 3 | 937 | ||
| AK093052.1 | - | 2 | 1255 | |||
| AK299344.1 | - | 3 | 1501 | |||
| BC034809.1 | - | 3 | 1452 | 4943538 | ||
| CX785083.1 | - | 1 | 416 | |||

FAM109A
Genomic Map