siRNA Details
| Gene Symbol: LACC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128303.1 | NP_001121775.1 | 4, 5 | 1394 | ||
| NM_153218.2 | NP_694950.2 | 4, 5 | 1230 | |||
| XM_005266261.3 | XP_005266318.1 | 4, 5 | 1853 | |||
| XM_006719766.3 | XP_006719829.1 | 4, 5 | 1011 | |||
| XM_006719767.3 | XP_006719830.1 | 4, 5 | 1230 | |||
| XM_011534933.2 | XP_011533235.1 | 5, 6 | 2048 | |||
| XM_011534934.2 | XP_011533236.1 | 4, 5 | 1167 | |||
| XM_017020394.1 | XP_016875883.1 | 4, 5 | 1287 | |||
| GenBank mRNA | AK096044.1 | - | 4, 5 | 1230 | ||
| BC035749.1 | - | 4, 5 | 1199 | 5750179 | ||

LACC1
Genomic Map