siRNA Details
| Gene Symbol: PHC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004427.3 | NP_004418.2 | 6 | 969 | ||
| NM_198040.2 | NP_932157.1 | 13 | 2276 | |||
| XM_005270570.1 | XP_005270627.1 | 6 | 905 | |||
| XM_017000513.1 | XP_016856002.1 | 14 | 2775 | |||
| XM_017000514.1 | XP_016856003.1 | 14 | 2387 | |||
| XM_017000515.1 | XP_016856004.1 | 14 | 2480 | |||
| XM_017000516.1 | XP_016856005.1 | 14 | 2709 | |||
| XM_017000517.1 | XP_016856006.1 | 14 | 2321 | |||
| XM_017000518.1 | XP_016856007.1 | 14 | 2414 | |||
| XM_017000519.1 | XP_016856008.1 | 14 | 2784 | |||
| XM_017000520.1 | XP_016856009.1 | 13 | 2234 | |||
| XM_017000521.1 | XP_016856010.1 | 13 | 2653 | |||
| GenBank mRNA | AJ419231.1 | - | 13 | 2276 | ||
| AK056429.1 | - | 7 | 1206 | |||
| AK128821.1 | - | 6 | 787 | |||
| AK292905.1 | - | 14 | 2714 | |||
| AK294426.1 | - | 8 | 1554 | |||
| BC015450.1 | - | 2 | 116 | |||
| BC018602.1 | - | 6 | 975 | |||
| BC028396.1 | - | 6 | 940 | |||
| BC029269.1 | - | 6 | 811 | |||
| BC068573.1 | - | 6 | 863 | |||
| BC092492.1 | - | 6 | 863 | |||
| BC110863.1 | - | 6 | 862 | |||
| BC130630.1 | - | 12 | 2192 | 40146661 | ||
| BC144121.1 | - | 12 | 2192 | 9052641 | ||
| U89278.1 | - | 6 | 958 | |||

PHC2
Genomic Map