siRNA Details
| Gene Symbol: LOC145783 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015419.2 | 1 | 933 | |||
| GenBank mRNA | AK056606.1 | - | 1 | 82 | ||
| AK096657.1 | - | 1 | 934 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC145783| Gene Symbol: LOC145783 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015419.2 | 1 | 933 | |||
| GenBank mRNA | AK056606.1 | - | 1 | 82 | ||
| AK096657.1 | - | 1 | 934 | |||