siRNA Details
| Gene Symbol: PGPEP1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001102612.2 | NP_001096082.2 | 5 | 683 | ||
| NM_001167902.1 | NP_001161374.1 | 5 | 1027 | |||
| XM_011521254.1 | XP_011519556.1 | 6 | 722 | |||
| XM_017021933.1 | XP_016877422.1 | 4 | 684 | |||
| GenBank mRNA | AK058162.1 | - | 5 | 1027 | ||
| BC104743.1 | - | 5 | 589 | |||
| BC104744.1 | - | 4 | 496 | |||
| BC105686.1 | - | 4 | 492 | |||

PGPEP1L
Genomic Map