siRNA Details
| Gene Symbol: TMCO5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152453.3 | NP_689666.2 | 12 | 893 | ||
| XM_005254168.3 | XP_005254225.1 | 12 | 874 | |||
| GenBank mRNA | BC029221.2 | - | 11 | 806 | 5167874 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMCO5A| Gene Symbol: TMCO5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152453.3 | NP_689666.2 | 12 | 893 | ||
| XM_005254168.3 | XP_005254225.1 | 12 | 874 | |||
| GenBank mRNA | BC029221.2 | - | 11 | 806 | 5167874 | |