siRNA Details
| Gene Symbol: OTOA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161683.1 | NP_001155155.1 | 11 | 1510 | ||
| NM_144672.3 | NP_653273.3 | 15 | 1654 | |||
| NM_170664.2 | NP_733764.1 | 6 | 748 | |||
| XM_011545747.2 | XP_011544049.1 | 16 | 2033 | |||
| XM_011545748.2 | XP_011544050.2 | 5 | 1027 | |||
| XM_017022950.1 | XP_016878439.1 | 16 | 1757 | |||
| GenBank mRNA | AK093062.1 | - | 6 | 748 | ||
| AK125840.1 | - | 11 | 1510 | |||
| AK302733.1 | - | 5 | 810 | |||
| BC129992.1 | - | 15 | 1651 | |||
| BC129993.1 | - | 15 | 1658 | 40128985 | ||

OTOA
Genomic Map