siRNA Details
| Gene Symbol: CARMIL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013838.1 | NP_001013860.1 | 32 | 3756 | ||
| NM_001317026.1 | NP_001303955.1 | 33 | 3648 | |||
| XM_011522874.1 | XP_011521176.1 | 33 | 3786 | |||
| XM_011522875.2 | XP_011521177.2 | 33 | 3786 | |||
| XM_017022953.1 | XP_016878442.1 | 33 | 3786 | |||
| GenBank mRNA | AB113647.1 | - | 32 | 3756 | ||
| BC043154.1 | - | 4 | 714 | |||
| FJ026014.1 | - | 33 | 3528 | |||

CARMIL2
Genomic Map