siRNA Details
| Gene Symbol: GSG1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109763.1 | NP_001103233.1 | 5 | 881 | ||
| NM_001323900.1 | NP_001310829.1 | 5 | 881 | |||
| NM_001323901.1 | NP_001310830.1 | 4 | 728 | |||
| NM_144675.2 | NP_653276.1 | 4 | 497 | |||
| XM_011545750.2 | XP_011544052.1 | 5 | 875 | |||
| GenBank mRNA | AK128775.1 | - | 5 | 502 | ||
| AY302134.1 | - | 3 | 331 | |||
| AY358206.1 | - | 4 | 720 | |||
| BC016460.1 | - | 4 | 497 | 4150450 | ||
| BI752847.1 | - | 6 | 704 | |||
| CV024949.1 | - | 3 | 331 | |||

GSG1L
Genomic Map