siRNA Details
| Gene Symbol: ZNF597 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152457.2 | NP_689670.1 | 4 | 1040 | ||
| GenBank mRNA | AK057633.1 | - | 4 | 1005 | ||
| BC029899.1 | - | 4 | 993 | 5175734 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF597| Gene Symbol: ZNF597 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152457.2 | NP_689670.1 | 4 | 1040 | ||
| GenBank mRNA | AK057633.1 | - | 4 | 1005 | ||
| BC029899.1 | - | 4 | 993 | 5175734 | ||