siRNA Details
| Gene Symbol: ZNF688 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024683.1 | NP_001019854.1 | 2, 3 | 409 | ||
| NM_145271.3 | NP_660314.1 | 2, 3 | 1097 | |||
| XM_005255139.2 | XP_005255196.1 | 2, 3 | 326 | |||
| XM_005255140.2 | XP_005255197.1 | 2, 3 | 946 | |||
| GenBank mRNA | AK122680.1 | - | 2, 3 | 1425 | ||
| BC018997.2 | - | 2, 3 | 376 | 4336167 | ||
| BM011208.1 | - | 2, 3 | 372 | |||
| KU178870.1 | - | 2, 3 | 319 | |||
| KU178871.1 | - | 1, 2 | 629 | |||

ZNF688
Genomic Map