siRNA Details
| Gene Symbol: ANKRD29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308238.1 | NP_001295167.1 | 9 | 910 | ||
| NM_173505.3 | NP_775776.2 | 10 | 1009 | |||
| XM_011525830.2 | XP_011524132.1 | 9 | 929 | |||
| XM_011525831.2 | XP_011524133.1 | 9 | 2949 | |||
| XM_011525832.2 | XP_011524134.1 | 9 | 3067 | |||
| XM_017025561.1 | XP_016881050.1 | 8 | 833 | |||
| GenBank mRNA | AK057782.1 | - | 9 | 840 | ||
| AK123197.1 | - | 7 | 2329 | |||
| AK313667.1 | - | 10 | 927 | |||
| BC030622.1 | - | 10 | 1014 | 4818619 | ||

ANKRD29
Genomic Map