siRNA Details
| Gene Symbol: C19orf84 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193623.1 | NP_001180552.1 | 2 | 834 | ||
| NR_126530.1 | 2 | 963 | ||||
| GenBank mRNA | BC043532.1 | - | 2 | 834 | ||
| BC085012.1 | - | 2 | 807 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C19orf84| Gene Symbol: C19orf84 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193623.1 | NP_001180552.1 | 2 | 834 | ||
| NR_126530.1 | 2 | 963 | ||||
| GenBank mRNA | BC043532.1 | - | 2 | 834 | ||
| BC085012.1 | - | 2 | 807 | |||