siRNA Details
| Gene Symbol: C19orf18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152474.4 | NP_689687.1 | 5 | 538 | ||
| GenBank mRNA | BC033933.2 | - | 5 | 541 | 5295864 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

C19orf18| Gene Symbol: C19orf18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152474.4 | NP_689687.1 | 5 | 538 | ||
| GenBank mRNA | BC033933.2 | - | 5 | 541 | 5295864 | |