siRNA Details
| Gene Symbol: HIPK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144685.4 | NP_653286.2 | 1 | 578 | ||
| XM_006723036.2 | XP_006723099.1 | 1 | 553 | |||
| GenBank mRNA | AK057380.1 | - | 1 | 553 | ||
| AK292228.1 | - | 1 | 539 | |||
| AK301713.1 | - | 2 | 331 | |||
| BC034501.2 | - | 1 | 544 | 4823304 | ||

HIPK4
Genomic Map