siRNA Details
| Gene Symbol: TMC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145303.2 | NP_001138775.2 | 6 | 1081 | ||
| NM_144686.3 | NP_653287.2 | 6 | 1063 | |||
| GenBank mRNA | AK314973.1 | - | 6 | 984 | ||
| AY236492.1 | - | 6 | 1080 | |||
| BC025323.1 | - | 6 | 970 | 2959382 | ||

TMC4
Genomic Map