siRNA Details
Gene Symbol: ZNF784 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203374.1 | NP_976308.1 | 2 | 1602 | ||
GenBank mRNA | AK074859.1 | - | 2 | 1593 | ||
BC065822.1 | - | 2 | 1602 | 6185645 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ZNF784 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203374.1 | NP_976308.1 | 2 | 1602 | ||
GenBank mRNA | AK074859.1 | - | 2 | 1593 | ||
BC065822.1 | - | 2 | 1602 | 6185645 |