siRNA Details
| Gene Symbol: ZNF563 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145276.2 | NP_660319.1 | 4 | 1479 | ||
| XM_005259750.4 | XP_005259807.1 | 4 | 956 | |||
| XM_005259751.3 | XP_005259808.1 | 5 | 1361 | |||
| XM_006722650.3 | XP_006722713.1 | 3 | 1416 | |||
| XM_006722651.3 | XP_006722714.1 | 4 | 1309 | |||
| XM_011527698.1 | XP_011526000.1 | 4 | 1299 | |||
| XM_011527699.2 | XP_011526001.1 | 3 | 1483 | |||
| XM_011527700.2 | XP_011526002.1 | 3 | 1254 | |||
| XM_017026332.1 | XP_016881821.1 | 4 | 1097 | |||
| GenBank mRNA | AK313755.1 | - | 4 | 1383 | ||
| BC022523.1 | - | 4 | 1447 | 4794529 | ||

ZNF563
Genomic Map