siRNA Details
| Gene Symbol: SPC24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317033.1 | NP_001303962.1 | 5 | 702 | ||
| NM_182513.3 | NP_872319.1 | 5 | 589 | |||
| XM_005259753.3 | XP_005259810.1 | 5 | 636 | |||
| GenBank mRNA | AK075287.1 | - | 5 | 559 | ||
| AK303157.1 | - | 5 | 446 | |||
| AK303266.1 | - | 5 | 697 | |||
| AY456387.1 | - | 5 | 553 | |||
| BC105037.1 | - | 5 | 585 | 8144040 | ||
| BC105039.1 | - | 5 | 585 | 8144042 | ||

SPC24
Genomic Map