siRNA Details
| Gene Symbol: SPC24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317032.1 | NP_001303961.1 | 4 | 465 | ||
| NM_001317033.1 | NP_001303962.1 | 4 | 578 | |||
| NM_182513.3 | NP_872319.1 | 4 | 465 | |||
| XM_005259753.3 | XP_005259810.1 | 4 | 512 | |||
| GenBank mRNA | AK075287.1 | - | 4 | 435 | ||
| AK303157.1 | - | 4 | 322 | |||
| AK303266.1 | - | 4 | 573 | |||
| AY456387.1 | - | 4 | 429 | |||
| BC105037.1 | - | 4 | 461 | 8144040 | ||
| BC105039.1 | - | 4 | 461 | 8144042 | ||
| BQ061822.1 | - | 4 | 422 | |||

SPC24
Genomic Map