siRNA Details
| Gene Symbol: DACT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301046.1 | NP_001287975.1 | 3 | 1574 | ||
| NM_145056.2 | NP_659493.2 | 4 | 1882 | |||
| XM_011526499.2 | XP_011524801.1 | 4 | 2658 | |||
| XM_011526500.2 | XP_011524802.1 | 3 | 1574 | |||
| XM_017026334.1 | XP_016881823.1 | 4 | 1743 | |||
| GenBank mRNA | BC016161.1 | - | 3 | 1573 | 2967514 | |

DACT3
Genomic Map