siRNA Details
| Gene Symbol: LGALS16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190441.2 | NP_001177370.2 | 3 | 298 | ||
| GenBank mRNA | BX095861.1 | - | 3 | 258 | ||
| FJ613352.1 | - | 3 | 298 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LGALS16| Gene Symbol: LGALS16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190441.2 | NP_001177370.2 | 3 | 298 | ||
| GenBank mRNA | BX095861.1 | - | 3 | 258 | ||
| FJ613352.1 | - | 3 | 298 | |||