siRNA Details
Gene Symbol: LGALS16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190441.2 | NP_001177370.2 | 2, 3 | 157 | ||
GenBank mRNA | BX095861.1 | - | 2, 3 | 117 | ||
FJ613352.1 | - | 2, 3 | 157 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LGALS16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190441.2 | NP_001177370.2 | 2, 3 | 157 | ||
GenBank mRNA | BX095861.1 | - | 2, 3 | 117 | ||
FJ613352.1 | - | 2, 3 | 157 |