siRNA Details
| Gene Symbol: ZNF599 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007248.2 | NP_001007249.1 | 4 | 930 | ||
| XM_017026369.1 | XP_016881858.1 | 4 | 1005 | |||
| GenBank mRNA | AK055225.1 | - | 4 | 916 | ||
| BC044615.1 | - | 5 | 1046 | 5266705 | ||

ZNF599
Genomic Map