siRNA Details
| Gene Symbol: PROSER3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039887.2 | NP_001034976.2 | 11 | 1523 | ||
| XM_011526533.2 | XP_011524835.1 | 11 | 2266 | |||
| XM_011526534.1 | XP_011524836.1 | 11 | 2266 | |||
| GenBank mRNA | AK055219.1 | - | 10 | 2292 | ||
| BC110893.1 | - | 10 | 1454 | 3914279 | ||

PROSER3
Genomic Map