siRNA Details
| Gene Symbol: PROSER3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039887.2 | NP_001034976.2 | 11 | 2090 | ||
| XM_011526528.1 | XP_011524830.1 | 11 | 2638 | |||
| XM_011526529.2 | XP_011524831.1 | 11 | 2649 | |||
| XM_011526530.1 | XP_011524832.1 | 11 | 2617 | |||
| XM_011526531.1 | XP_011524833.1 | 11 | 2605 | |||
| XM_011526532.1 | XP_011524834.1 | 11 | 2472 | |||
| XM_011526533.2 | XP_011524835.1 | 11 | 2833 | |||
| XM_011526534.1 | XP_011524836.1 | 12 | 2638 | |||
| XM_011526535.2 | XP_011524837.1 | 9 | 2345 | |||
| GenBank mRNA | AK055219.1 | - | 10 | 2859 | ||
| BC110893.1 | - | 10 | 2020 | 3914279 | ||

PROSER3
Genomic Map