siRNA Details
| Gene Symbol: ZNF98 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098626.1 | NP_001092096.1 | 4 | 641 | ||
| XM_011527705.1 | XP_011526007.1 | 4 | 600 | |||
| GenBank mRNA | BC110576.1 | - | 3 | 431 | 40007052 | |
| BC129810.1 | - | 3 | 431 | |||

ZNF98
Genomic Map