siRNA Details
| Gene Symbol: ZNF98 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098626.1 | NP_001092096.1 | 4 | 670 | ||
| XM_011527705.1 | XP_011526007.1 | 4 | 629 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF98| Gene Symbol: ZNF98 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098626.1 | NP_001092096.1 | 4 | 670 | ||
| XM_011527705.1 | XP_011526007.1 | 4 | 629 | |||