siRNA Details
| Gene Symbol: ZNF714 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182515.3 | NP_872321.2 | 5 | 649 | ||
| NR_117086.1 | 6 | 668 | ||||
| NR_117087.1 | 5 | 649 | ||||
| GenBank mRNA | AK056006.1 | - | 5 | 598 | ||
| BC022527.1 | - | 6 | 612 | |||
| BC037782.1 | - | 6 | 617 | 4797729 | ||
| BC050468.2 | - | 5 | 589 | |||
| BC064985.1 | - | 5 | 524 | |||
| BM788748.1 | - | 3 | 384 | |||

ZNF714
Genomic Map