siRNA Details
| Gene Symbol: EFNB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004093.3 | NP_004084.1 | 4 | 655 | ||
| XM_017020406.1 | XP_016875895.1 | 4 | 8916 | |||
| GenBank mRNA | BC069342.1 | - | 4 | 582 | 7262269 | |
| BC074856.2 | - | 4 | 582 | 30915225 | ||
| BC074857.2 | - | 4 | 582 | 30915462 | ||
| BC105955.1 | - | 4 | 600 | 40034079 | ||
| BC105956.1 | - | 4 | 600 | 40034078 | ||
| BC105957.1 | - | 4 | 600 | 40034080 | ||
| L38734.1 | - | 4 | 610 | |||
| U16797.1 | - | 4 | 586 | |||
| U81262.1 | - | 4 | 586 | |||

EFNB2
Genomic Map