siRNA Details
| Gene Symbol: DIRAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145173.3 | NP_660156.1 | 2 | 687 | ||
| XM_011527713.2 | XP_011526015.1 | 2 | 852 | |||
| GenBank mRNA | AB076888.1 | - | 1 | 628 | ||
| AY056037.1 | - | 1 | 529 | |||
| AY059641.1 | - | 1 | 529 | |||
| AY180973.1 | - | 1 | 529 | |||
| BC030660.1 | - | 2 | 666 | 4814337 | ||

DIRAS1
Genomic Map