siRNA Details
| Gene Symbol: ZNF555 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172775.1 | NP_001166246.1 | 4 | 573 | ||
| NM_152791.4 | NP_690004.4 | 4 | 576 | |||
| XM_011527716.2 | XP_011526018.1 | 4 | 576 | |||
| XM_017026375.1 | XP_016881864.1 | 4 | 576 | |||
| GenBank mRNA | AK093163.1 | - | 4 | 561 | ||
| AK292954.1 | - | 4 | 521 | |||
| AL832140.1 | - | 4 | 537 | |||
| BC022022.2 | - | 4 | 527 | 4822197 | ||
| BX647423.1 | - | 4 | 534 | |||

ZNF555
Genomic Map