siRNA Details
Gene Symbol: C1orf127 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170754.1 | NP_001164225.1 | 9 | 894 | ||
XM_017000373.1 | XP_016855862.1 | 7 | 746 | |||
GenBank mRNA | AK094437.1 | - | 4 | 398 | ||
BC126349.1 | - | 5 | 484 | 8992065 | ||
BC143794.1 | - | 5 | 484 | 9052308 | ||
BC143795.1 | - | 5 | 484 | 9052309 | ||
BC143797.1 | - | 5 | 484 | 9052311 |
