siRNA Details
| Gene Symbol: PHF13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153812.2 | NP_722519.2 | 4 | 1161 | ||
| XM_011540762.1 | XP_011539064.1 | 4 | 963 | |||
| GenBank mRNA | AB209544.1 | - | 4 | 853 | ||
| AK027492.1 | - | 3 | 686 | |||
| AK097715.1 | - | 4 | 1095 | |||
| AK315110.1 | - | 4 | 1093 | |||
| AL121733.1 | - | 2 | 649 | |||
| AY069975.1 | - | 4 | 791 | |||
| BC032792.2 | - | 2 | 588 | |||
| BC038516.1 | - | 4 | 1172 | 5266750 | ||

PHF13
Genomic Map