siRNA Details
| Gene Symbol: PHF13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153812.2 | NP_722519.2 | 2 | 451 | ||
| XM_011540762.1 | XP_011539064.1 | 2 | 253 | |||
| GenBank mRNA | AB209544.1 | - | 2 | 143 | ||
| AK097715.1 | - | 2 | 385 | |||
| AK315110.1 | - | 2 | 383 | |||
| AY069975.1 | - | 2 | 81 | |||
| BC038516.1 | - | 2 | 462 | 5266750 | ||
| DB064673.1 | - | 3 | 386 | |||

PHF13
Genomic Map