siRNA Details
| Gene Symbol: MFSD4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181644.4 | NP_857595.3 | 2 | 474 | ||
| XM_011509229.2 | XP_011507531.1 | 2 | 474 | |||
| GenBank mRNA | AK091896.1 | - | 2 | 415 | ||
| AK293816.1 | - | 2 | 474 | |||
| AK295148.1 | - | 2 | 322 | |||
| AK304109.1 | - | 1 | 279 | |||
| AK304120.1 | - | 2 | 401 | |||
| BC036549.2 | - | 2 | 424 | 5264261 | ||
| DC317463.1 | - | 2 | 474 | |||

MFSD4A
Genomic Map