siRNA Details
| Gene Symbol: RC3H1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300850.1 | NP_001287779.1 | 11 | 2237 | ||
| NM_001300851.1 | NP_001287780.1 | 11 | 2237 | |||
| NM_001300852.1 | NP_001287781.1 | 11 | 2237 | |||
| NM_172071.3 | NP_742068.1 | 11 | 2237 | |||
| XM_005244921.3 | XP_005244978.1 | 11 | 2174 | |||
| XM_011509231.2 | XP_011507533.1 | 11 | 2237 | |||
| GenBank mRNA | AB095945.1 | - | 11 | 2078 | ||
| AK093501.1 | - | 2 | 386 | |||
| AK122948.1 | - | 11 | 2237 | |||
| BC136784.1 | - | 11 | 2191 | 9020774 | ||
| BC144408.1 | - | 11 | 2191 | 9052929 | ||

RC3H1
Genomic Map