siRNA Details
| Gene Symbol: ZNF362 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152493.2 | NP_689706.2 | 9 | 1811 | ||
| XM_005270507.2 | XP_005270564.1 | 8 | 1951 | |||
| XM_005270508.4 | XP_005270565.1 | 9 | 2608 | |||
| XM_017000415.1 | XP_016855904.1 | 8 | 2523 | |||
| GenBank mRNA | AF318356.1 | - | 8 | 1661 | ||
| AK098342.1 | - | 1 | 51 | |||
| BC071632.1 | - | 2 | 623 | |||
| BC103704.1 | - | 1 | 244 | |||
| BC104779.1 | - | 8 | 1733 | 8143782 | ||
| BC112113.1 | - | 8 | 1732 | 8327581 | ||

ZNF362
Genomic Map