siRNA Details
| Gene Symbol: DCST1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143687.2 | NP_001137159.1 | 4 | 358 | ||
| NM_152494.3 | NP_689707.2 | 5 | 433 | |||
| GenBank mRNA | AK057347.1 | - | 5 | 415 | ||
| AK301876.1 | - | 4 | 335 | |||
| AK301902.1 | - | 5 | 512 | |||
| BC064844.1 | - | 4 | 388 | 30378702 | ||

DCST1
Genomic Map