siRNA Details
| Gene Symbol: DCST1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152494.3 | NP_689707.2 | 4 | 355 | ||
| GenBank mRNA | AK057347.1 | - | 4 | 337 | ||
| AK301902.1 | - | 4 | 434 | |||
| BC064844.1 | - | 3 | 310 | 30378702 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DCST1| Gene Symbol: DCST1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152494.3 | NP_689707.2 | 4 | 355 | ||
| GenBank mRNA | AK057347.1 | - | 4 | 337 | ||
| AK301902.1 | - | 4 | 434 | |||
| BC064844.1 | - | 3 | 310 | 30378702 | ||