siRNA Details
| Gene Symbol: ZNF648 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017000302.1 | XP_016855791.1 | 4 | 3602 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF648| Gene Symbol: ZNF648 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017000302.1 | XP_016855791.1 | 4 | 3602 | ||