siRNA Details
| Gene Symbol: C1orf210 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164829.1 | NP_001158301.1 | 3 | 1351 | ||
| NM_182517.2 | NP_872323.1 | 3 | 1422 | |||
| XM_011540802.1 | XP_011539104.1 | 3 | 1481 | |||
| GenBank mRNA | BC041633.1 | - | 3 | 1412 | 4868093 | |

C1orf210
Genomic Map