siRNA Details
| Gene Symbol: FAM71A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153606.3 | NP_705834.2 | 1 | 1059 | ||
| GenBank mRNA | AK057358.1 | - | 1 | 672 | ||
| AK315021.1 | - | 1 | 683 | |||
| BC035007.1 | - | 1 | 1034 | 5267155 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM71A| Gene Symbol: FAM71A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153606.3 | NP_705834.2 | 1 | 1059 | ||
| GenBank mRNA | AK057358.1 | - | 1 | 672 | ||
| AK315021.1 | - | 1 | 683 | |||
| BC035007.1 | - | 1 | 1034 | 5267155 | ||