siRNA Details
| Gene Symbol: FAM71A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153606.3 | NP_705834.2 | 1 | 1559 | ||
| GenBank mRNA | AK057358.1 | - | 1 | 1172 | ||
| AK301981.1 | - | 2 | 805 | |||
| AK315021.1 | - | 1 | 1183 | |||
| BC035007.1 | - | 1 | 1534 | 5267155 | ||

FAM71A
Genomic Map