siRNA Details
| Gene Symbol: PRNT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024267.1 | 2 | 1032 | |||
| NR_024268.1 | 2 | 795 | ||||
| NR_024269.1 | 2 | 572 | ||||
| GenBank mRNA | AJ427539.1 | - | 2 | 796 | ||
| AJ427540.1 | - | 2 | 1032 | |||
| AL137296.1 | - | 2 | 572 | |||
| BC137389.1 | - | 1 | 449 | |||
| BC137390.1 | - | 1 | 450 | |||

PRNT
Genomic Map