siRNA Details
| Gene Symbol: LOC149950 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034152.1 | 3 | 485 | |||
| GenBank mRNA | BC111383.1 | - | 3 | 485 | 5169644 | |
| BC130462.1 | - | 2 | 283 | 40146493 | ||
| BC130464.1 | - | 2 | 283 | 40146495 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC149950| Gene Symbol: LOC149950 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034152.1 | 3 | 485 | |||
| GenBank mRNA | BC111383.1 | - | 3 | 485 | 5169644 | |
| BC130462.1 | - | 2 | 283 | 40146493 | ||
| BC130464.1 | - | 2 | 283 | 40146495 | ||