siRNA Details
Gene Symbol: LOC149950 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034152.1 | 3 | 804 | |||
GenBank mRNA | BC111383.1 | - | 3 | 804 | 5169644 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC149950 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034152.1 | 3 | 804 | |||
GenBank mRNA | BC111383.1 | - | 3 | 804 | 5169644 |